• USA Home
  • 10774 - Antimony(III) chloride

EMAIL THIS PAGE TO A FRIEND
10774 Sigma-Aldrich

Antimony(III) chloride

puriss. p.a., ACS reagent, ≥99.0% (RT)

Synonym: Antimony trichloride

Purchase

Properties

Related Categories A-B, Puriss p.a. ACS, Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Antimony Salts,
grade   ACS reagent
vapor density   7.9 (vs air)
vapor pressure   1 mmHg ( 49 °C)
InChI Key   FAPDDOBMIUGHIN-UHFFFAOYSA-K
grade   puriss. p.a.
assay   ≥99.0% (RT)
mp   73.4 °C (lit.)
anion traces   sulfate (SO42-): ≤50 mg/kg
cation traces   As: ≤200 mg/kg
  Ca: ≤50 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤20 mg/kg
  Zn: ≤5 mg/kg
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1733 8 / PGII
WGK Germany 
2
RTECS 
CC4900000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?