EMAIL THIS PAGE TO A FRIEND
45860 Sigma-Aldrich

Butyl acetate

puriss. p.a., ACS reagent

  • CAS Number 123-86-4

  • Linear Formula CH3COO(CH2)3CH3

  • Molecular Weight 116.16

  •  Beilstein/REAXYS Number 1741921

  •  EC Number 204-658-1

  •  MDL number MFCD00009445

  •  PubChem Substance ID 329756992

  •  NACRES NA.21

Purchase

Properties

Related Categories Alcohol Acetates, Biochemicals and Reagents, Cell Biology, Fatty Acyls, Fatty Esters,
Quality Level   200
grade   ACS reagent
vapor density   4 (vs air)
vapor pressure   15 mmHg ( 25 °C)
  8 mmHg ( 20 °C)
grade   puriss. p.a.
assay   ≥99.5% (GC)
form   liquid
autoignition temp.   790 °F
expl. lim.   7.6 %
impurities   substances darkened by H2SO4, in accordance
  ≤0.01% free acid (as CH3COOH)
  ≤0.1% butyl formate (GC)
  ≤0.1% butyl propionate (GC)
  ≤0.1% water
  ≤0.2% butyl alcohol (GC)
evapn. residue   ≤0.001%
refractive index   n20/D 1.394 (lit.)
  n20/D 1.394
pH   6.2 (20 °C, 5.3 g/L)
bp   124-126 °C (lit.)
mp   −78 °C (lit.)
density   0.88 g/mL at 25 °C (lit.)
cation traces   Al: ≤0.5 mg/kg
  B: ≤0.02 mg/kg
  Ba: ≤0.1 mg/kg
  Bi: ≤0.1 mg/kg
  Ca: ≤0.5 mg/kg
  Cd: ≤0.05 mg/kg
  Co: ≤0.02 mg/kg
  Cr: ≤0.02 mg/kg
  Cu: ≤0.02 mg/kg
  Fe: ≤0.1 mg/kg
  K: ≤0.5 mg/kg
  Li: ≤0.1 mg/kg
  Mg: ≤0.1 mg/kg
  Mn: ≤0.02 mg/kg
  Mo: ≤0.1 mg/kg
  Na: ≤0.5 mg/kg
  Ni: ≤0.02 mg/kg
  Pb: ≤0.1 mg/kg
  Sn: ≤0.1 mg/kg
  Sr: ≤0.1 mg/kg
  Zn: ≤0.1 mg/kg
SMILES string   CCCCOC(C)=O
InChI   1S/C6H12O2/c1-3-4-5-8-6(2)7/h3-5H2,1-2H3
InChI key   DKPFZGUDAPQIHT-UHFFFAOYSA-N

Description

General description

Butyl acetate is a butyl ester of acetic acid. It exists in four isomeric forms: butyl acetate, sec-butyl acetate, isobutyl acetate and tert-butyl acetate. It is a colorless pleasant smelling liquid. Its photocatalytic decomposition has been carried out in a fixed-bed continuous annular reactor in the presence of Pt/TiO2and WO3/TiO2 catalysts.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Central nervous system
Supplemental Hazard Statements 
EUH066
RIDADR 
UN1123 - class 3 - PG 3 - Butyl acetates
WGK Germany 
WGK 1
RTECS 
AF7350000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?