EMAIL THIS PAGE TO A FRIEND
50486 Sigma-Aldrich

Hydriodic acid

puriss. p.a., 55-60%, not stabilized, packed in coated, shock- and leak-protected glass bottle

Synonym: Hydriotic acid

Purchase

Properties

Related Categories Acids, Acids & Bases, Alternative Energy, Chemical Synthesis, Halide Sources,
InChI Key   XMBWDFGMSWQBCA-UHFFFAOYSA-N
grade   puriss. p.a.
assay   55-60%
quality   not stabilized
  packed in coated, shock- and leak-protected glass bottle
impurities   ≤0.001% phosphorous compounds (as PO4)
ign. residue   ≤0.005% (as SO4)
bp   127 °C (lit.)
density   1.7 g/mL at 20 °C
anion traces   bromide, chloride (as Cl-): ≤100 mg/kg
  sulfate (SO42-): ≤20 mg/kg
cation traces   Ag: ≤0.1 mg/kg
  Al: ≤1 mg/kg
  As: ≤1 mg/kg
  Ba: ≤0.1 mg/kg
  Bi: ≤0.1 mg/kg
  Ca: ≤1 mg/kg
  Cd: ≤0.1 mg/kg
  Co: ≤0.1 mg/kg
  Cr: ≤0.1 mg/kg
  Cu: ≤0.1 mg/kg
  Fe: ≤2 mg/kg
  K: ≤1 mg/kg
  Li: ≤0.1 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤0.1 mg/kg
  Mo: ≤0.1 mg/kg
  Na: ≤1 mg/kg
  Ni: ≤0.1 mg/kg
  Pb: ≤0.1 mg/kg
  Sr: ≤0.1 mg/kg
  Tl: ≤0.1 mg/kg
  Zn: ≤0.5 mg/kg

Description

Caution

may discolor to red-brown on storage

Other Notes

Sales restrictions may apply

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1787 8 / PGII
WGK Germany 
1
Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?