• USA Home
  • 63100 - Magnesium perchlorate hydrate

EMAIL THIS PAGE TO A FRIEND
63100 Sigma-Aldrich

Magnesium perchlorate hydrate

puriss. p.a., ≥99.0% (calc. based on dry substance, KT)

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Essential Chemicals, I-N, Puriss p.a.,
InChI Key   KKFFNQXCJMNXHI-UHFFFAOYSA-L
grade   puriss. p.a.
assay   ≥99.0% (calc. based on dry substance, KT)
impurities   ≤34% water
anion traces   chloride (Cl-): ≤50 mg/kg
  nitrate (NO3-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤500 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤100 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

General description

Magnesium perchlorate hydrate is a hydrated magnesium salt. Phases in the crystals of magnesium perchlorate hydrate system (Mg(ClO4)2·nH2O; n = 4,2) and their crystal structures have been investigated by X-ray powder diffraction (XRD) analyses.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1475 5.1 / PGII
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?