• USA Home
  • 09972 - 3-Nitrotetrazolium blue chloride

EMAIL THIS PAGE TO A FRIEND
09972 Sigma-Aldrich

3-Nitrotetrazolium blue chloride

≥75% (AT)

Synonym: 3,3′-(3,3′-Dimethoxy-4,4′-biphenylene)bis[5-(3-nitrophenyl)-2-phenyl-2H-tetrazolium chloride]

Purchase

Properties

Related Categories Alkaline Phosphatase, Biochemicals and Reagents, Cytokinin dehydrogenase, Enzyme Substrates, Enzymes, Inhibitors, and Substrates,
assay   ≥75% (AT)
form   powder
storage temp.   2-8°C
SMILES string   [Cl-].[Cl-].COc1cc(ccc1-n2nc(n[n+]2-c3ccccc3)-c4ccccc4)-c5ccc(c(OC)c5)-n6nc(n[n+]6-c7ccccc7)-c8ccccc8
InChI   1S/C40H32N8O2.2ClH/c1-49-37-27-31(23-25-35(37)47-43-39(29-15-7-3-8-16-29)41-45(47)33-19-11-5-12-20-33)32-24-26-36(38(28-32)50-2)48-44-40(30-17-9-4-10-18-30)42-46(48)34-21-13-6-14-22-34;;/h3-28H,1-2H3;2*1H/q+2;;/p-2
InChI key   MUUHXGOJWVMBDY-UHFFFAOYSA-L

Description

General description

rest mainly solvent (methanol or ethanol)

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?