• USA Home
  • 104100 - Sigma 104® Phosphatase Substrate

EMAIL THIS PAGE TO A FRIEND
104100 Sigma-Aldrich

Sigma 104® Phosphatase Substrate

weight 100 mg (capsule)

Synonym: 4-Nitrophenyl phosphate disodium salt hexahydrate, p-Nitrophenyl phosphate disodium hexahydrate, Disodium 4-nitrophenyl phosphate hexahydrate, di-Sodium 4-nitrophenyl phosphate, pNPP disodium hexahydrate

  • CAS Number 333338-18-4

  • Linear Formula O2NC6H4OP(O)(ONa)2 · 6H2O

  • Molecular Weight 371.14

  •  Beilstein/REAXYS Number 5324661

  •  EC Number 224-246-5

  •  MDL number MFCD00066288

Purchase

Properties

InChI Key   KAKKHKRHCKCAGH-UHFFFAOYSA-L
clinical   for in vitro diagnostic use
IVD   for in vitro diagnostic use
weight   100 mg (capsule)
storage temp.   −20°C

Description

Application

Chromogenic substrate for the determination of acid and alkaline phosphatases. The reaction produces a soluble end product that is yellow in color. The reaction may be stopped with 3 N NaOH and read at 405 nm.

Legal Information

Sigma 104 is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?