• USA Home
  • 11448 - Nonyl-β-D-glucoside 50 mM Solution

EMAIL THIS PAGE TO A FRIEND
11448 Sigma-Aldrich

Nonyl-β-D-glucoside 50 mM Solution

50 mM

Synonym: Detergent Screening Solution 06/Kit-No 66317, Nonyl β-D-glucopyranoside solution

Purchase

Properties

InChI Key   QFAPUKLCALRPLH-UXXRCYHCSA-N
concentration   50 mM
storage temp.   2-8°C
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?