• USA Home
  • 11548 - 5-Azido-2-nitrobenzoic acid

EMAIL THIS PAGE TO A FRIEND
11548 Sigma

5-Azido-2-nitrobenzoic acid

≥98.0% (HPLC)

Synonym: 2-Nitro-5-azidobenzoic acid

  • CAS Number 60117-34-2

  • Empirical Formula (Hill Notation) C7H4N4O4

  • Molecular Weight 208.13

  •  Beilstein/REAXYS Number 1884110

  •  MDL number MFCD00043015

  •  PubChem Substance ID 24847219

Purchase

Properties

assay   ≥98.0% (HPLC)
mp   163-165 °C
storage temp.   2-8°C

Description

Other Notes

Reagent for introducing a photoaffinity label in peptides and proteins.

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1325 - class 4.1 - PG 2 - Flammable solids, organic, n.o.s., HI: all
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?