• USA Home
  • 1503 - Sodium chloride solution

EMAIL THIS PAGE TO A FRIEND
1503 Sigma

Sodium chloride solution

0.15 M

Synonym: Isotonic saline

  • CAS Number 7647-14-5

  • Linear Formula NaCl

  • Molecular Weight 58.44

  •  Beilstein/REAXYS Number 3534976

  •  MDL number MFCD00003477

Purchase

Properties

InChI Key   FAPWRFPIFSIZLT-UHFFFAOYSA-M
clinical   for in vitro diagnostic use
concentration   0.15 M
storage temp.   2-8°C
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
2

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?