• USA Home
  • 1570L - BIOEAZE® Storage and Transport

EMAIL THIS PAGE TO A FRIEND
1570L SAFC

BIOEAZE® Storage and Transport

Purchase

Properties

description   Tank Liner, Pillow Style; Tank Dimension 18" diameter x 33" height (130L)
packaging   pkg of 20 pkg
size   130 L

Description

General description

29" diameter x 48" height

Legal Information

BIOEAZE is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?