EMAIL THIS PAGE TO A FRIEND
16707 Sigma-Aldrich

ADREPMA®

for chromatography, A-20

Purchase

Properties

grade   for chromatography
mean particle size   20 μm
density   1.10-1.14 g/mL at 20 °C
storage temp.   −20°C

Description

General description

Silica-based aligned protein A affinity chromatography phase with high dynamic binding capacity, low leaching enabling speed and long life
Particle size: 20 micrometer
Elution of human IgG at pH 4.5

Legal Information

ADREPMA is a registered trademark of Osaka Soda Co., Ltd.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?