• USA Home
  • 181C - β-Glucuronidase, Lymphocyte

EMAIL THIS PAGE TO A FRIEND
181C Sigma-Aldrich

β-Glucuronidase, Lymphocyte

Purchase

Properties

Related Categories Cell Biology, Cytochemistry Products, Hematology and Histology More...
Quality Level   GMP-IVD
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use
shipped in   wet ice
storage temp.   2-8°C

Description

Diagnostic Implications

Demonstration of lymphocyte β-glucuronidase activity (focal staining pattern) in cytocentrifuge or buffy coat preparations from blood, bone marrow films and tissue touch preparations typifies T-lymphocytes.

Kit component also available separately

Description

Product #

Add to Cart

Acetate Solution (386-3) 50 mL    
Citrate Solution (91-5) 50 mL    
Methylene Blue Solution (180-8) 50 mL    
Pararosaniline Solution (180-4) 5 mL    

Kit component only

Description

Product #

Add to Cart

Naphthol AS-BI β-D-Glucuronic Acid solution (kit only) 50 mL    
Sodium Nitrite (kit only) 10 tablets    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin


Milli-Q® Water Purification Solutions

The Emprove® Program
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

1804 Pararosaniline solution
915 Citrate Solution, pH 3.6±0.1 (25 °C), 27 mM
3863 Acetate solution, 2.5 M, pH 5.2
1808 Methylene Blue solution, 1.4 % (w/v) in 95% ethanol
R8382 Reagent Alcohol, used for histology tissue preparation

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?