• USA Home
  • 208S - GH Positive Control Slides

EMAIL THIS PAGE TO A FRIEND
208S Sigma-Aldrich

GH Positive Control Slides

suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections)

  •  NACRES NA.24

Purchase

Properties

Related Categories Antibodies, Cell Marque Control Slides, Cell Marque Products for Pathology More...
Quality Level   100
  500
packaging   pkg of 5 slides
mfr. no.   Cell Marque
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
shipped in   ambient
storage temp.   2-8°C

Description

General description

GH positive control slides are intended for use as positive controls for immunohistochemical (IHC) staining using an anti-GH antibody.

Physical form

Paraffin embedded tissue section mounted on microscope slide

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

antibody

Product #

Description

Add to Cart

208A-7 GH Rabbit Polyclonal Antibody

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?