• USA Home
  • 24581C - EX-CELL® MDCK Serum-Free Medium for MDCK Cells

EMAIL THIS PAGE TO A FRIEND
24581C SAFC

EX-CELL® MDCK Serum-Free Medium for MDCK Cells

Animal-protein free, with L-glutamine, without sodium bicarbonate, dry powder, suitable for cell culture

Purchase

Properties

Related Categories CHO Media and Medium Optimization Kits, CHO Platform, Cell Culture, Serum-free Media
Quality Level   500
description   for research or for further manufacturing use
form   dry powder
availability   not available in Japan
application(s)   cell culture | mammalian: suitable
storage temp.   2-8°C

Description

Application

EX-CELL MDCK is an animal-protein free, serum-free medium developed for the long-term growth of Madin Darby Canine Kidney (MDCK) and related cells. The cells, in an attachment culture, can be subcultured directly into EX-CELL MDCK from serum-supplemented media without adaptation. Cell densities and doubling times achieved under serum-free conditions are comparable to those achieved in a serum-supplemented culture.

Quantity

Formulated to contain 21.6 grams of powder per liter of medium.

Legal Information

EX-CELL is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Pipette Controller from Brand
Protocols & Articles

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Coronavirus COVID-19 (SARS-CoV-2) Detection, Characterization & Vaccine Production

As a global life science tools and equipment supplier, we are here to help in the response to COVID-19. From supplying raw materials and research products for detection and characterization to produc...
Keywords: PAGE

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?