• USA Home
  • 289A-7 - Myeloperoxidase Rabbit Polyclonal Antibody

EMAIL THIS PAGE TO A FRIEND
289A-7 Sigma-Aldrich

Myeloperoxidase Rabbit Polyclonal Antibody

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Cell Marque Antibodies, Hematopathology, MUE-MZ,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500
species reactivity   human
form   buffered aqueous solution
shipped in   wet ice
storage temp.   2-8°C
packaging   vial of 0.1 mL concentrate (289A-74)
  vial of 0.5 mL concentrate (289A-75)
  bottle of 1.0 mL predilute (289A-77)
  vial of 1.0 mL concentrate (289A-76)
  bottle of 7.0 mL predilute (289A-78)
antibody form   Ig fraction of antiserum
antibody product type   primary antibodies
description   For In Vitro Diagnostic Use in Select Regions (See Chart)
mfr. no.   Cell Marque
concentration   1.0-10.0 μg/mL (predilute)
  100-1000 μg/mL (concentrate)
control   bone marrow
visualization   cytoplasmic

Description

General description

Anti-myeloperoxidase detects granulocytes and monocytes in blood and precursors of granulocytes in the bone marrow. This antibody can detect myeloid cell populations of the bone marrow as well as in other sites.

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Legal Information

Cell Marque is a trademark of Sigma-Aldrich Co. LLC

Quality


IVD

IVD

IVD

RUO

Linkage

Myeloperoxidase Positive Control Slides , Product No. 289S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Download the latest released IFU
Note: This IFU may not apply to your specific product lot.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

control

Product #

Description

Add to Cart

289S Myeloperoxidase Positive Control Slides, suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections) 

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?