• USA Home
  • 297127 - Atrazine-ring-UL-14C

EMAIL THIS PAGE TO A FRIEND
297127 Sigma-Aldrich

Atrazine-ring-UL-14C

≥95% (HPLC), solid

Synonym: 2-chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine-ring-ul-14C

Purchase

Properties

assay   ≥95% (HPLC)
form   solid
extent of labeling   5-25 mCi per mmol
mass shift   M+6
storage temp.   2-8°C
Gene Information   mouse ... Esr1(13982)

Description

Packaging

Combi-vial

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?