• USA Home
  • 297178 - Benzoic acid-ring-UL-14C

EMAIL THIS PAGE TO A FRIEND
297178 Sigma-Aldrich

Benzoic acid-ring-UL-14C

toluene solution

Purchase

Properties

form   toluene solution
extent of labeling   1-15 mCi per mmol
mass shift   M+12
storage temp.   2-8°C

Description

Packaging

sealed ampule

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?