• USA Home
  • 297453 - Propionic acid-1-14C sodium salt

EMAIL THIS PAGE TO A FRIEND
297453 Sigma-Aldrich

Propionic acid-1-14C sodium salt

aqueous solution

Purchase

Properties

form   aqueous solution
extent of labeling   1-10 mCi per mmol
mass shift   M+2
storage temp.   2-8°C

Description

Packaging

sealed ampule

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?