• USA Home
  • 304697 - Dodecanoic acid-(carboxy-14C)

EMAIL THIS PAGE TO A FRIEND
304697 Sigma-Aldrich

Dodecanoic acid-(carboxy-14C)

≥98% (Radiochemical Purity, HPLC), solid

Synonym: Dodecanoic-1-14C acid

Purchase

Properties

assay   ≥98% (Radiochemical Purity, HPLC)
form   solid
extent of labeling   5-25 mCi per mmol
mass shift   M+2
storage temp.   2-8°C
SMILES string   CCCCCCCCCCC[14C](O)=O

Description

Packaging

Combi-vial

Safety & Documentation

Safety Information

RIDADR 
UN2910 Radioactive material, excepted package-limited quantity of material
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?