• USA Home
  • 30957 - 2-Deoxy-D-ribono-1,4-lactone

EMAIL THIS PAGE TO A FRIEND
30957 Sigma-Aldrich

2-Deoxy-D-ribono-1,4-lactone

≥95% (GC)

Synonym: (4S,5R)-4-Hydroxy-5-(hydroxymethyl)dihydrofuran-2(3H)-one, 2-Deoxy-D-ribonic acid γ-lactone

  • CAS Number 34371-14-7

  • Empirical Formula (Hill Notation) C5H8O4

  • Molecular Weight 132.11

  •  Beilstein/REAXYS Number 81260

  •  MDL number MFCD15144952

  •  PubChem Substance ID 329753608

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates D-F, Monosaccharide More...
InChI Key   YIXDEYPPAGPYDP-IUYQGCFVSA-N
assay   ≥95% (GC)
optical activity   [α]/D 19.0±2.5°, c = 1 in H2O
suitability   conforms to structure for Proton NMR spectrum
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?