EMAIL THIS PAGE TO A FRIEND
313114 Sigma-Aldrich

Formaldehyde-14C

aqueous solution

Purchase

Properties

form   aqueous solution
extent of labeling   5-55 mCi per mmol
mass shift   M+2
SMILES string   [H][14C]([H])=O

Description

Packaging

sealed ampule

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3
Flash Point(F) 
147 °F
Flash Point(C) 
64 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?