• USA Home
  • 313718 - 1-Naphthylacetic acid-car­boxy-14C

EMAIL THIS PAGE TO A FRIEND
313718 Sigma-Aldrich

1-Naphthylacetic acid-car­boxy-14C

solid

Synonym: 1-Naphthaleneacetic-car­boxy-14C acid

Purchase

Properties

form   solid
extent of labeling   1-15 mCi per mmol
storage temp.   2-8°C

Description

Packaging

screw cap bottle

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?