EMAIL THIS PAGE TO A FRIEND
313912 Sigma-Aldrich

Oxalic acid-14C

solid

Purchase

Properties

form   solid
extent of labeling   1-10 mCi per mmol
mass shift   M+4
storage temp.   2-8°C
SMILES string   O[14C](=O)[14C](O)=O

Description

Packaging

screw cap bottle

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?