• USA Home
  • 313920 - Palmitic-(carboxy-14C) acid

EMAIL THIS PAGE TO A FRIEND
313920 Sigma-Aldrich

Palmitic-(carboxy-14C) acid

solid

  • CAS Number 765-07-1

  • Linear Formula CH3[CH2]1414COOH

  • Molecular Weight 256.42

  •  MDL number MFCD00055822

Purchase

Properties

form   solid
extent of labeling   1-15 mCi per mmol
mass shift   M+2
storage temp.   2-8°C

Description

Packaging

screw cap bottle

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?