• USA Home
  • 359920 - 2-(4-Biphenylyl)-prop-2-yl 4′-methoxycarbonylphenyl carbonate

EMAIL THIS PAGE TO A FRIEND
359920 Sigma-Aldrich

2-(4-Biphenylyl)-prop-2-yl 4′-methoxycarbonylphenyl carbonate

~97% (HPLC)

Synonym: BPOC reagent

  • CAS Number 31140-37-1

  • Empirical Formula (Hill Notation) C24H22O5

  • Molecular Weight 390.43

  •  Beilstein/REAXYS Number 2179522

  •  EC Number 250-482-3

  •  MDL number MFCD00075450

Purchase

Properties

assay   ~97% (HPLC)
mp   90-100 °C (dec.)
storage temp.   2-8°C
SMILES string   COC(=O)c1ccc(OC(=O)OC(C)(C)c2ccc(cc2)-c3ccccc3)cc1
InChI   1S/C24H22O5/c1-24(2,20-13-9-18(10-14-20)17-7-5-4-6-8-17)29-23(26)28-21-15-11-19(12-16-21)22(25)27-3/h4-16H,1-3H3
InChI key   PXYPBZYXNPAGFW-UHFFFAOYSA-N

Description

Application

Reagent for introduction of acid-labile amine-protecting group BPOC.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?