• USA Home
  • 36495 - 4-[(1-Naphtho[2,3-d]triazol-1-yl)methyl]benzoic acid

EMAIL THIS PAGE TO A FRIEND
36495 Sigma-Aldrich

4-[(1-Naphtho[2,3-d]triazol-1-yl)methyl]benzoic acid

for fluorescence, ≥98.0% (HPLC)

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Indicators and Probes, Fluorescent Probes, Labels, Particles and Stains, Nitric Oxide Probes, Nitric Oxide and Oxygen Probes More...
grade   for fluorescence
assay   ≥98.0% (HPLC)
suitability   in accordance for fluorescence
SMILES string   OC(=O)c1ccc(Cn2nnc3cc4ccccc4cc23)cc1
InChI   1S/C18H13N3O2/c22-18(23)13-7-5-12(6-8-13)11-21-17-10-15-4-2-1-3-14(15)9-16(17)19-20-21/h1-10H,11H2,(H,22,23)
InChI key   CPVINIGXGKJNOS-UHFFFAOYSA-N

Description

Other Notes

Reference probe for the cell-permeable, non-fluorescent NO probe Ethyl 4-[(3-amino-2-naphthyl)aminomethyl]benzoate hydrochloride (Cat. No. 16513).

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Sensor Dyes

In 1998 R.F. Furchgott, L.J. Ignarro, and F. Murad received the Nobel Prize in Physiology and Medicine for their discoveries concerning nitric oxide as a signaling molecule in the cardiovascular syst...
Monika Baeumle
BioFiles 2011, 6.3, 23.
Keywords: Applications, Biochemistry, Biological processes, Cancer, Cardiovascular, Central Nervous System, Diseases, Extinction coefficient, Indicators, Inflammation, Methods, Microscopy, Neurotransmission, Sol-gel, Type

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?