• USA Home
  • 38463 - 1-[2-(Maleimido)ethyl]-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium triflate

EMAIL THIS PAGE TO A FRIEND
38463 SAFC

1-[2-(Maleimido)ethyl]-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium triflate

BioReagent, suitable for fluorescence, ≥90% (HPCE)

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains, Other Fluorescent Labels
InChI Key   LVIALPCMAFGHPU-UHFFFAOYSA-M
product line   BioReagent
assay   ≥90% (HPCE)
fluorescence   λex 397 nm; λem 569 nm in 0.1 M phosphate pH 7.0
suitability   suitable for fluorescence
storage temp.   2-8°C

Description

General description

Highly photostable, water soluble fluorescent label with large Stoke′s shift, high QY in aqueous media and pH tolerance. Major disadvantage: rel. low extinction coefficient (epsilon slightly >20′000 in EtOH).

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?