• USA Home
  • 39197 - N-Dansylethylenediamine

EMAIL THIS PAGE TO A FRIEND
39197 Sigma-Aldrich

N-Dansylethylenediamine

suitable for fluorescence

Synonym: N-(2-Aminoethyl)-5-dimethylamino-1-naphthalenesulfonamide, N-(2-Aminoethyl)dansylamide, DNSEDA, NSC 666752

  • CAS Number 35060-08-3

  • Empirical Formula (Hill Notation) C14H19N3O2S

  • Molecular Weight 293.38

  •  Beilstein/REAXYS Number 2817535

  •  MDL number MFCD00467523

  •  PubChem Substance ID 329755827

  •  NACRES NA.32

Purchase

Properties

Related Categories Biochemicals and Reagents, Luminescent Compounds/Detection, Stains and Dyes More...
Quality Level   100
assay   ≥90% (TLC)
solubility   methanol: 1 mg/mL, clear, slightly yellow to deep yellow
fluorescence   λex 360 nm; λem 530 nm±10 nm in methanol
suitability   suitable for fluorescence
storage temp.   −20°C
SMILES string   CN(C)c1cccc2c(cccc12)S(=O)(=O)NCCN
InChI   1S/C14H19N3O2S/c1-17(2)13-7-3-6-12-11(13)5-4-8-14(12)20(18,19)16-10-9-15/h3-8,16H,9-10,15H2,1-2H3
InChI key   CSJXLKVNKAXFSI-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?