• USA Home
  • 41965 - Ampholyte high resolution

EMAIL THIS PAGE TO A FRIEND
41965 Sigma

Ampholyte high resolution

pH 2.0-4.0, ~40% in H2O

Synonym: SinuLyte

Purchase

Description

General description

This series of carrier ampholytes is based on a specific method to produce mixtures of numerous polyamino-polycarboxylic acids. These carrier ampholytes have been developed for use of isoelectric focusing in quality control applications, especially in food and seed analysis.
• High buffer capacity: Compared to established ampholytes they offer buffering capacities 1.5-2.5 times higher (up to pH 8, equal above pH 8)
• Good, even conductivity
• Linear and reproducible pH gradient
• High resolution

Legal Information

SinuLyte is a trademark of Sinus Biochemistry and Electrophoresis GmbH

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?