• USA Home
  • 42914 - Pinosylvin monomethyl ether

EMAIL THIS PAGE TO A FRIEND
42914 Sigma-Aldrich

Pinosylvin monomethyl ether

≥97.0% (HPLC)

Synonym: (E)-3-Hydroxy-5-methoxystilbene, (E)-3-Methoxy-5-(2-phenylethenyl)phenol, 5-Methoxy-3-stilbenol, Pinosylvin 3-methyl ether

  • CAS Number 35302-70-6

  • Empirical Formula (Hill Notation) C15H14O2

  • Molecular Weight 226.27

  •  Beilstein/REAXYS Number 1875807

  •  MDL number MFCD00210546

  •  PubChem Substance ID 329756282

Purchase

Properties

assay   ≥97.0% (HPLC)
mp   118-122 °C
SMILES string   COc1cc(O)cc(\C=C\c2ccccc2)c1
InChI   1S/C15H14O2/c1-17-15-10-13(9-14(16)11-15)8-7-12-5-3-2-4-6-12/h2-11,16H,1H3/b8-7+
InChI key   JVIXPWIEOVZVJC-BQYQJAHWSA-N

Description

Application

Pinosylvin, a pre-infectious stilbenoid toxin, is used to study its properties as a fungitoxin and potential therapeutic agent. Pinosylvin monomethyl ether, a methylated derivative of pinosylvin, may be used as a reference material in the analysis of stibenoid toxins. Pinosylvin monomethyl ether may also be used as a representative stilbene to study its biological actions and therapeutic value in processes such as cell survival, apoptosis and cell mobility.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?