• USA Home
  • 43096 - 1,2-Dimyristoyl-sn-glycero-3-phospho-rac-(1-glycerol) ammonium salt

EMAIL THIS PAGE TO A FRIEND
43096 Sigma-Aldrich

1,2-Dimyristoyl-sn-glycero-3-phospho-rac-(1-glycerol) ammonium salt

≥97.0% (TLC)

Synonym: 1,2-Dimyristoyl-sn-glycero-3-phosphoglycerol ammonium salt, 1,2-Ditetradecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) ammonium salt, L-α-Phosphatidyl-DL-glycerol, dimyristoyl ammonium salt, DMPG-NH4, PG(14:0/14:0), Tetradecanoic acid 1,1′-[(1R)-1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl] ester monoammonium salt

  • CAS Number 953758-30-0

  • Empirical Formula (Hill Notation) C34H70NO10P

  • Molecular Weight 683.89

  •  PubChem Substance ID 329756305

Purchase

Properties

Related Categories Cell Biology, Glycerophospholipid Metabolism, Metabolic Pathways, Metabolomics
Quality Level   100
biological source   synthetic
InChI Key   XDGIUHZTOUFLGK-SKZICHJRSA-N
assay   ≥97.0% (TLC)
storage temp.   −20°C

Description

Application

Has been used as the neutral lipid in studies of highly defined mixed-lipid liposome encapsulated drugs, e.g., a platinum complex antitumor agent, the antifungal agent hamycin, and Candida albicans ribosomes as an anti-Candida vaccine.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?