EMAIL THIS PAGE TO A FRIEND
43378 Sigma-Aldrich

Atto 390 alkyne

BioReagent, suitable for fluorescence

  •  NACRES NA.32

Purchase

Properties

Related Categories Atto Labels, Biochemicals and Reagents, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains
product line   BioReagent
fluorescence   λex 390 nm; λem 479 nm±10 nm in 0.1 M phosphate pH 7.0
suitability   suitable for fluorescence
storage temp.   −20°C

Description

General description

Atto 390 is a fluorescent label with a coumarin structure. The dye is intended for application in the area of life science, e.g. labeling of DNA, RNA or proteins. Characteristic features of the label are high fluorescence quantum yield, large Stokes-shift, good photostability and low molecular weight.
The alkyne modification is suitable for reactions with azide groups (Huisgen reaction - “Click Chemistry“).

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?