EMAIL THIS PAGE TO A FRIEND
45725 Sigma-Aldrich

Acetic acid

for luminescence, BioUltra

Synonym: Glacial acetic acid

Purchase

Properties

grade   for luminescence
vapor density   2.07 (vs air)
product line   BioUltra
autoignition temp.   800 °F
expl. lim.   16 %, 92 °F
  4 %, 59 °F
impurities   insoluble matter, passes filter test
evapn. residue   ≤0.0005%
refractive index   n20/D 1.371 (lit.)
bp   117-118 °C (lit.)
mp   16.2 °C (lit.)
density   1.049 g/mL at 25 °C (lit.)
anion traces   chloride (Cl-): ≤0.5 mg/kg
  phosphate (PO43-): ≤0.5 mg/kg
  sulfate (SO42-): ≤0.5 mg/kg
cation traces   Ag: ≤0.01 mg/kg
  Al: ≤0.05 mg/kg
  Ba: ≤0.01 mg/kg
  Be: ≤0.01 mg/kg
  Bi: ≤0.1 mg/kg
  Ca: ≤0.2 mg/kg
  Cd: ≤0.02 mg/kg
  Co: ≤0.01 mg/kg
  Cr: ≤0.05 mg/kg
  Cu: ≤0.01 mg/kg
  Fe: ≤0.2 mg/kg
  Ge: ≤0.05 mg/kg
  K: ≤0.1 mg/kg
  Li: ≤0.01 mg/kg
  Mg: ≤0.1 mg/kg
  Mn: ≤0.01 mg/kg
  Mo: ≤0.02 mg/kg
  Na: ≤0.5 mg/kg
  Ni: ≤0.05 mg/kg
  Pb: ≤0.02 mg/kg
  Sr: ≤0.01 mg/kg
  Ti: ≤0.01 mg/kg
  Tl: ≤0.05 mg/kg
  V: ≤0.01 mg/kg
  Zn: ≤0.05 mg/kg
  Zr: ≤0.1 mg/kg
λ   neat
SMILES string   CC(O)=O
UV absorption   λ: 260 nm Amax: ≤0.1
  λ: 280 nm Amax: ≤0.02
suitability   in accordance for luminescence
InChI   1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)
InChI key   QTBSBXVTEAMEQO-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2789PSN1 3(8) / PGII
WGK Germany 
1
RTECS 
AF1225000
Flash Point(F) 
104 °F
Flash Point(C) 
40 °C
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?