• USA Home
  • 50447 - (±)-1,3-Butanediol 4 M solution

EMAIL THIS PAGE TO A FRIEND
50447 Sigma-Aldrich

(±)-1,3-Butanediol 4 M solution

Synonym: (±)-1,3-Butanediol solution, Additive Screening Solution 03/Kit-No 78374

Purchase

Properties

storage temp.   2-8°C
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?