• USA Home
  • 53601 - Anti-Rabbit IgG−Abberior® CAGE 532 antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
53601 Sigma-Aldrich

Anti-Rabbit IgG−Abberior® CAGE 532 antibody produced in goat

for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)

Synonym: Abberior® CAGE 532-Anti-Rabbit IgG antibody produced in goat

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, RAB-RAD
antibody product type   primary antibodies
fluorescence   λex 530 nm; λem 550 nm±10 nm in PBS
storage temp.   −20°C

Description

Analysis Note

May contain significant amounts of BSA as stabilizer

unconjugated dye≤5% of total fluorescence

General description

Absorption Maximum, λmax: 237, 302, 350 nm (non-activated, PBS, pH 7.4)
533 nm (photoactivated, PBS, pH 7.4)
Extinction Coefficient, ε(λmax): 82′000 M-1cm-1 (photoactivated, PBS, pH 7.4)
Fluorescence Maximum, λfl: 541 nm (PBS, pH 7.4)
Photoactivation Wavelength: 360 - 440 nm
Recommended STED Wavelength, λSTED: 610 - 640 nm
Fluorescence Quantum Yield, η: 0.84 (after photoactivation, PBS, pH 7.4)
Fluorescence Lifetime, τ: 4.1 (PBS, pH 7.4)

Other Notes

Find more infomation here Abberior®

Legal Information

abberior is a registered trademark of Abberior GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary and Secondary Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay needs. Use the Antibody...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

95705 Abberior® CAGE 532, maleimide, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
52996 Anti-Mouse IgG−Abberior® CAGE 532 antibody produced in goat, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?