• USA Home
  • 60352 - Potassium phosphate dibasic

EMAIL THIS PAGE TO A FRIEND
60352 Sigma-Aldrich

Potassium phosphate dibasic

BioUltra, for luminescence, anhydrous, ≥99.0% (T)

Synonym: Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate

Purchase

Properties

grade   for luminescence
InChI Key   ZPWVASYFFYYZEW-UHFFFAOYSA-L
product line   BioUltra
assay   ≥99.0% (T)
impurities   insoluble matter, passes filter test
pH   8.7-9.3 (25 °C, 1 M in H2O)
solubility   H2O: 1 M at 20 °C, clear, colorless
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤10 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Na: ≤5000 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Zn: ≤5 mg/kg
λ   1 M in H2O
UV absorption   λ: 260 nm Amax: ≤0.20
  λ: 280 nm Amax: ≤0.15
suitability   in accordance for luminescence
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?