• USA Home
  • 617-GS - Outer Root Sheath Cell Growth Supplements (5 ml-4 parts)

EMAIL THIS PAGE TO A FRIEND
617-GS Sigma-Aldrich

Outer Root Sheath Cell Growth Supplements (5 ml-4 parts)

  •  NACRES NA.71

Purchase

Properties

Related Categories Cell Culture, Hair Cell Media, Media, Primary Cell Culture
form   liquid
shelf life   1 yr
shipped in   wet ice
storage temp.   2-8°C

Description

General description

Added to Basal Medium to create Growth Medium

Quality

Each lot was tested for the ability to support plating, spreading and proliferation

Preparation Note

contained in protocol

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Pipette Controller from Brand
Protocols & Articles

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Introduction to Primary Cell Culture [VIDEO]

Interested in receiving more content like this? Join over 8,000 of your peers who have already signed up for the Cell Informer newsletter.
Keywords: Cell culture, Drug discovery, Pharmaceutical

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?