• USA Home
  • 62278 - Lipase, immobilized in Sol-Gel-AK from Candida rugosa

EMAIL THIS PAGE TO A FRIEND
62278 Sigma-Aldrich

Lipase, immobilized in Sol-Gel-AK from Candida rugosa

≥10 U/g

Purchase

Description

Application

Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

Packaging

1 g in glass bottle

Biochem/physiol Actions

Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol oleic acid per minute at pH 7.5 and 37 °C (triolein, Cat. No. 62314 as substrate)

Immobilized lipase with excellent characteristics

Other Notes

Immobilized lipase with excellent characteristics

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
RTECS 
TO9776500
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?