EMAIL THIS PAGE TO A FRIEND
64670 Sigma-Aldrich

Methocel® 65 HG

27-29% methoxyl basis

Synonym: Methyl cellulose

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Core Bioreagents, Polysaccharide, Research Essentials More...
assay   27-29% methoxyl basis
concentration   5.5-7.5% hydroxypropyl
viscosity   3000-5600 mPa.s, 2 % in H2O(20 °C)
SMILES string   [*]OC[C@H]1O[C@@H](O[C@@H]2[C@@H](CO[*])O[C@@H](O[*])[C@H](O[*])[C@H]2O[*])[C@H](O[*])[C@@H](O[*])[C@@H]1O[*]
InChI key   YLGXILFCIXHCMC-JHGZEJCSSA-N

Description

Legal Information

Methocel is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?