• USA Home
  • 68397 - Trehalose 6-octanoate

EMAIL THIS PAGE TO A FRIEND
68397 Sigma-Aldrich

Trehalose 6-octanoate

≥95.0% (GC)

Synonym: α,α-Trehalose 6-caprylate, α-D-Glucopyranosyl-α-D-glucopyranoside 6-octanoate, Trehalose 6-caprylate

  • CAS Number 64622-90-8

  • Empirical Formula (Hill Notation) C20H36O12

  • Molecular Weight 468.49

  •  PubChem Substance ID 329760885

Purchase

Properties

InChI Key   WNPJPTWSRIRPIJ-LPKQXWMESA-N
assay   ≥95.0% (GC)
CMC   5.6 mM
storage temp.   2-8°C

Description

Application

Trehalose is commonly used as a stabilizer and protective material for biomaterial compounds. Because of these characteristics, Trehalose is introduced as a hydrophilic part. Due to these properties of Trehalose, protein degeneration is expected to be prevented with Trehalose detergents. Therefore, Trehalose detergents may keep the function of the protein and be utilized in the field of proteomics research.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?