• USA Home
  • 68835 - 1-Methylpiperidine solution

EMAIL THIS PAGE TO A FRIEND
68835 Sigma-Aldrich

1-Methylpiperidine solution

BioReagent, suitable for protein sequencing

Synonym: N-Methylpiperidine

  • CAS Number 626-67-5

  • Empirical Formula (Hill Notation) C6H13N

  • Molecular Weight 99.17

  •  Beilstein/REAXYS Number 1560282

  •  MDL number MFCD00006491

  •  PubChem Substance ID 24885853

Purchase

Properties

product line   BioReagent
impurities   ≥20% methanol
suitability   suitable for protein sequencing
storage temp.   −20°C
SMILES string   CN1CCCCC1
InChI   1S/C6H13N/c1-7-5-3-2-4-6-7/h2-6H2,1H3
InChI key   PAMIQIKDUOTOBW-UHFFFAOYSA-N

Description

Packaging

Special packaging for direct use in common sequencers.

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN 3286 8(6.1)(3) / PGII
WGK Germany 
2
Flash Point(F) 
41 °F
Flash Point(C) 
5 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?