• USA Home
  • 68891 - 1,4-Butanediol 4 M solution

EMAIL THIS PAGE TO A FRIEND
68891 Sigma-Aldrich

1,4-Butanediol 4 M solution

Synonym: 1,4-Butanediol solution, Additive Screening Solution 04/Kit-No 78374

Purchase

Properties

InChI Key   WERYXYBDKMZEQL-UHFFFAOYSA-N
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?