• USA Home
  • 70478 - Naphthol AS-TR phosphate

EMAIL THIS PAGE TO A FRIEND
70478 Sigma-Aldrich

Naphthol AS-TR phosphate

for histology, ≥98.0% (HPLC)

  • CAS Number 2616-72-0

  • Empirical Formula (Hill Notation) C18H15ClNO5P

  • Molecular Weight 391.74

  •  Beilstein/REAXYS Number 6666070

  •  EC Number 220-046-7

  •  MDL number MFCD00042716

  •  PubChem Substance ID 57652251

Purchase

Properties

Related Categories Acid Phosphatase, Alkaline Phosphatase, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates,
grade   for histology
assay   ≥98.0% (HPLC)
form   powder
storage temp.   2-8°C
SMILES string   Cc1cc(Cl)ccc1NC(=O)c2cc3ccccc3cc2OP(O)(O)=O
InChI   1S/C18H15ClNO5P/c1-11-8-14(19)6-7-16(11)20-18(21)15-9-12-4-2-3-5-13(12)10-17(15)25-26(22,23)24/h2-10H,1H3,(H,20,21)(H2,22,23,24)
InChI key   SHTFMUPUBYXPCD-UHFFFAOYSA-N

Description

Application

Substrate for the histochemical demonstration of acid and alkaline phosphatase.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?