• USA Home
  • 70760 - 2-Amino-1,5-naphthalenedisulfonic acid

EMAIL THIS PAGE TO A FRIEND
70760 Sigma

2-Amino-1,5-naphthalenedisulfonic acid

Synonym: 2-Naphthylamine-1,5-disulfonic acid

  • CAS Number 117-62-4

  • Empirical Formula (Hill Notation) C10H9NO6S2

  • Molecular Weight 303.31

  •  Beilstein/REAXYS Number 2888645

  •  EC Number 204-201-6

  •  MDL number MFCD00021512

  •  PubChem Substance ID 24886125

Purchase

Properties

impurities   ~17% water

Description

Analysis Note

may contain NaCl

Other Notes

Automated sequential degradation of non-polar peptides

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
QJ6135000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?