EMAIL THIS PAGE TO A FRIEND
75975 Sigma

Page blue G 90

for microscopy

Synonym: Brilliant blue G, Brilliant indocyanine G

  • CAS Number 6104-58-1

  • Empirical Formula (Hill Notation) C47H48N3NaO7S2

  • Molecular Weight 854.02

  •  Colour Index Number 42655

  •  EC Number 228-058-4

  •  MDL number MFCD00078482

  •  PubChem Substance ID 57652598

Purchase

Properties

grade   for microscopy
InChI Key   RWVGQQGBQSJDQV-UHFFFAOYSA-M
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?