• USA Home
  • 76439 - Pentaethylene glycol monohexadecyl ether

EMAIL THIS PAGE TO A FRIEND
76439 Sigma-Aldrich

Pentaethylene glycol monohexadecyl ether

BioXtra, ≥99.0% (TLC)

Synonym: C16E5, Hexadecyl pentaethylene glycol ether, Hexadecylpentaglycol, Polyoxyethylene (5) cetyl ether

  • CAS Number 4478-97-1

  • Linear Formula CH3(CH2)15(OCH2CH2)5OH

  • Molecular Weight 462.70

  •  Beilstein/REAXYS Number 2287681

  •  MDL number MFCD00043365

  •  PubChem Substance ID 57652638

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents N to Z, Non-Ionic Detergents
description   non-ionic
product line   BioXtra
assay   ≥99.0% (TLC)
mol wt   462.70 g/mol
application(s)   protein purification: suitable
mp   39-40 °C
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cr: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤50 mg/kg
  Mg: ≤50 mg/kg
  Mn: ≤50 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg
SMILES string   CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCO
InChI   1S/C26H54O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-17-28-19-21-30-23-25-32-26-24-31-22-20-29-18-16-27/h27H,2-26H2,1H3
InChI key   CJZQCJWPIYNMQG-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?