EMAIL THIS PAGE TO A FRIEND
76663 Supelco

ZnAF-1

≥95.0% (HPCE)

Synonym: 5-{2-[Bis(2-pyridylmethyl)amino]ethylamino}fluorescein

Purchase

Properties

Related Categories Biochemicals and Reagents, Chelators and Ion Probes, Fluorescent Indicators and Probes, Fluorescent Probes, Labels, Particles and Stains, Zinc Probes More...
Quality Level   100
assay   ≥95.0% (HPCE)
fluorescence   λex 492 nm; λem 514 nm in PBS, Zn2+
SMILES string   OC(=O)c1cc(NCCN(Cc2ccccn2)Cc3ccccn3)ccc1C4=C5C=CC(=O)C=C5Oc6cc(O)ccc46
InChI   1S/C34H28N4O5/c39-25-8-11-28-31(18-25)43-32-19-26(40)9-12-29(32)33(28)27-10-7-22(17-30(27)34(41)42)37-15-16-38(20-23-5-1-3-13-35-23)21-24-6-2-4-14-36-24/h1-14,17-19,37,39H,15-16,20-21H2,(H,41,42)
InChI key   SAVLVCGHDLKAAF-UHFFFAOYSA-N

Description

Application

suitable for the fluorimetric detection of Zn2+

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?