• USA Home
  • 77431 - Phe-Ala-Ala-Phe(4-NO2)-Phe-Val-Leu (4-pyridylmethyl) ester

EMAIL THIS PAGE TO A FRIEND
77431 Sigma-Aldrich

Phe-Ala-Ala-Phe(4-NO2)-Phe-Val-Leu (4-pyridylmethyl) ester

≥94% (HPLC)

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience, Enzyme Substrates, Enzymes, Inhibitors, and Substrates,
assay   ≥94% (HPLC)
form   powder
storage temp.   −20°C
SMILES string   CC(C)C[C@H](NC(=O)[C@@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](Cc2ccc(cc2)[N+]([O-])=O)NC(=O)[C@H](C)NC(=O)[C@H](C)NC(=O)[C@@H](N)Cc3ccccc3)C(C)C)C(=O)OCc4ccncc4
InChI   1S/C50H63N9O10/c1-30(2)25-42(50(66)69-29-37-21-23-52-24-22-37)57-49(65)43(31(3)4)58-48(64)41(27-35-15-11-8-12-16-35)56-47(63)40(28-36-17-19-38(20-18-36)59(67)68)55-45(61)33(6)53-44(60)32(5)54-46(62)39(51)26-34-13-9-7-10-14-34/h7-24,30-33,39-43H,25-29,51H2,1-6H3,(H,53,60)(H,54,62)(H,55,61)(H,56,63)(H,57,65)(H,58,64)/t32-,33-,39-,40-,41-,42-,43-/m0/s1
InChI key   GHSCHCKVTVGANO-RQHOAPLWSA-N

Description

Application

suitable as substrate for pepsin

Other Notes

Substrate for a fast continuous spectrophotometric assay of cathepsin D. Substrate for pepsin

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?