EMAIL THIS PAGE TO A FRIEND
77723 Sigma-Aldrich

Pefachrome® Try

≥95.0% (HPLC)

Synonym: Bz-Val-Gly-Arg-4-nitroanilide acetate, N-Benzoyl-L-valyl-glycyl-L-arginine-4-nitroanilide acetate

  • CAS Number 161572-28-7

  • Empirical Formula (Hill Notation) C26H34N8O6 · C2H4O2

  • Molecular Weight 614.65

  •  MDL number MFCD03791098

  •  PubChem Substance ID 57652708

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Substrates by Enzyme, Trypsin Substrates More...
Quality Level   100
assay   ≥95.0% (HPLC)
form   powder
solubility   0.05 M phosphate, pH 7.6: 5 mg/mL, clear, colorless
storage temp.   2-8°C
SMILES string   CC(O)=O.CC(C)[C@H](NC(=O)c1ccccc1)C(=O)NCC(=O)N[C@@H](CCCNC(N)=N)C(=O)Nc2ccc(cc2)[N+]([O-])=O
InChI   1S/C26H34N8O6.C2H4O2/c1-16(2)22(33-23(36)17-7-4-3-5-8-17)25(38)30-15-21(35)32-20(9-6-14-29-26(27)28)24(37)31-18-10-12-19(13-11-18)34(39)40;1-2(3)4/h3-5,7-8,10-13,16,20,22H,6,9,14-15H2,1-2H3,(H,30,38)(H,31,37)(H,32,35)(H,33,36)(H4,27,28,29);1H3,(H,3,4)/t20-,22-;/m0./s1
InChI key   JWECNHBSLLUDLK-DTRWSJPISA-N

Description

Application

Pefachrome® substrates are used for chromogenic assays of various serine proteinases. Pefachrome® Try is used for the detection of serine proteases such as trypsin.

Packaging

1 piece contains 10 μmol Pefachrome® Try and 20 mg glycine

Legal Information

Pefachrome is a registered trademark of Pentapharm AG, Basel

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?