EMAIL THIS PAGE TO A FRIEND
81462 Sigma-Aldrich

Ponceau S solution

for electrophoresis, ~0.1 % (w/v) Ponceau S in 5% acetic acid

  • CAS Number 6226-79-5

  • Empirical Formula (Hill Notation) C22H12N4Na4O13S4

  • Molecular Weight 760.57

  •  MDL number MFCD00003892

  •  PubChem Substance ID 24887781

Purchase

Properties

grade   for electrophoresis
InChI Key   KQHKSGRIBYJYFX-WNDMICSESA-J
concentration   ~0.1 % (w/v) Ponceau S in 5% acetic acid
refractive index   n20/D 1.338
density   1.01 g/mL at 20 °C
suitability   in accordance for electrophoresis test
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?